• USA Home
  • T2031 - Ultramicro pipette tips
EMAIL THIS PAGE TO A FRIEND
T2031 Sigma
Ultramicro pipette tips
Microcapillary, round tip, volume range 10 μL, non-sterile
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Miscellaneous Pipette Tips, Pipette Tips, Pipettes, Pipettors and Tips More...
material   polypropylene
  colorless tip
sterility   non-sterile
feature   barrier: no
packaging   tray of 10 × 20 tips
mfr. no.   Ultramicro
volume range   10 μL
Description
Application
For loading tube gels and slab gels 0.6 mm or thicker.
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?