• USA Home
  • T2276 - cis-9, trans-11-Tetradecadienyl acetate

EMAIL THIS PAGE TO A FRIEND
T2276 Sigma

cis-9, trans-11-Tetradecadienyl acetate

~95%

Synonym: (Z,E)-9,11-TDDA

Purchase

Properties

Related Categories Alcohol Acetates, Biochemicals and Reagents, Esters, Fatty Acids and conjugates, Fatty Acyls,
assay   ~95%
bp   85-86 °C/0.003 mmHg(lit.)
storage temp.   −20°C

Description

Biochem/physiol Actions

Pheromone for Spodoptera littoralis and Spodoptera litura.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
141.8 °F
Flash Point(C) 
61 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?