• USA Home
  • T2409 - Tubing cartridges for multichannel pump head
EMAIL THIS PAGE TO A FRIEND
T2409 Sigma
Tubing cartridges for multichannel pump head
large
Purchase
Properties
feature   flow rate 0.8 ‑ 3.8 mL per revolution using 1/8′′ to 5/16′′ I.D. tubing
mfr. no.   Scilog
availability   not available in EU (Not available in EU countries)
  not available in EU
Description
Compatibility
Accepts tubing from 1/4 to 7/16 in. (6.4 to 11.1 mm) O.D.; 1/16 in. (1.6 mm) wall is recommended for all tubing sizes.
Features and Benefits
Easy-loading design with tubing retainers on both sides. Snap locks allow one-hand installation on the pump head without tools. Occlusion is separately adjustable in each cartridge for fine control of flow rates.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?