• USA Home
  • T2414 - Silicone rubber laboratory tubing
EMAIL THIS PAGE TO A FRIEND
T2414 Sigma
Silicone rubber laboratory tubing
I.D. × O.D. 1/8 in. × 5/16 in.
Purchase
Properties
Related Categories Labware, Silicone Tubing, Tubing More...
I.D. × O.D.   1/8 in. × 5/16 in.
wall size   3/32 in.
Description
General description
Translucent and flexible. Temp. range: -60 to 460°F (-51 to 238°C). Do not use with concentrated acids or dilute sodium hydroxide. Suitable for high-temperature applications.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
T2289 Silicone rubber laboratory tubing, I.D. × O.D. 1/8 in. × 1/4 in.
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?