• USA Home
  • T2416 - Barnant hand-held thermometer
EMAIL THIS PAGE TO A FRIEND
T2416 Sigma
Barnant hand-held thermometer
Model 115
Purchase
Properties
mfr. no.   Barnant
measuring range   temperature, accuracy: 0.4 °C, (throughout range)
battery power   9 V
Description
Compatibility
Accepts J, K, T, and E thermocouples.
Features and Benefits
Features eight-reading recall, scanning for maximum or minimum temperature, δT, freeze display, field calibration, and automatic compensation for inherent probe error.
General description
All-solid state, microprocessor controlled meters accept a wide variety of thermocouple junction types and offer unmatched flexibility, range and accuracy.
• 4-digit LCD with selectable resolution (0.1° or 1°), reading °C or °F (auto-select above 1000°)
• Accepts ANSI standard thermocouple probes with mini-connectors (see Probes for specific ranges)
• Calibrated against NBS curves for each of the thermocouple types
• Built-in tilt stand for desktop use
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?