• USA Home
  • T2443 - Greiner centrifuge tubes

EMAIL THIS PAGE TO A FRIEND
T2443 Sigma

Greiner centrifuge tubes

centrifuge tube, 50 mL, 30 x 115 mm, conical (V) bottom, w/ graduations, I.D. field, w/ rack

Synonym: Greiner centrifuge tubes, centrifuge tubes

Purchase

Properties

Related Categories 50 mL centrifuge tubes, Centrifugation Supplies, Centrifuge Tubes, Plastic, Greiner, Greiner Centrifuge Tubes,
material   polypropylene tube
sterility   sterile
feature   graduations, ID field, w/ rack
mfr. no.   Greiner 227270
packaging   pack of 25
  case of 300
capacity   50 mL
diam. × L   30 mm × 115 mm
color   blue cap
  colorless tube

Description

General description

Greiner centrifuge tubes
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.

Price and Availability


Steriflip Filters Units
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?