• USA Home
  • T2516 - Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato) ytterbium(III) derivative

EMAIL THIS PAGE TO A FRIEND
T2516 Sigma

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato) ytterbium(III) derivative

Synonym: Yb[fod]3

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

225835 Resolve-Al YbFOD

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?