• USA Home
  • T2538 - Trizma® hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
T2538 Sigma

Trizma® hydrochloride solution

pH 7.2, 1 M

Synonym: Tris hydrochloride solution

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions, Buffers A to Z More...
grade   for molecular biology
InChI Key   QKNYBSVHEMOAJP-UHFFFAOYSA-N
sterility   0.2 μm filtered
concentration   1 M
impurities   DNase, RNase, NICKase and protease, none detected
pH   7.2
Featured Industry   Diagnostic Assay Manufacturing

Description

Application

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per degree C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Other Notes

Prepared with pH-adjusted Biotechnology Performance Certified Trizma Base in 18 megohm water and 0.2 μm filtered.

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?