• USA Home
  • T2568 - Anti-TRPC4 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
T2568 Sigma

Anti-TRPC4 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym: Anti-Transient receptor potential cation channel, subfamily C, member 4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Calcium Channels, Antibodies to Ion Channels,
species reactivity   mouse (predicted to detect), rat
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Trpc4(22066)
rat ... Trpc4(84494)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide (KEKHAHEEDSSIDYDL) corresponding to residues 943-958 of mouse TRPC4 with additional N-terminal cysteine.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?