• USA Home
  • T2777 - Anti-3,3′,5-Triiodo-L-Thyronine (T3) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
T2777 Sigma

Anti-3,3′,5-Triiodo-L-Thyronine (T3) antibody produced in rabbit

whole antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Hormones, Antibodies to Other Protein/Peptide Hormones,
biological source   rabbit
antibody form   whole antiserum
clone   polyclonal
contains   15 mM sodium azide
application(s)   radioimmunoassay: 1:6,000
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

Secondary antibody-polyethylene glycol 125I RIA

Immunogen

T3-BSA

Reconstitution

Dilute with buffer: 0.01 M phosphate, pH 7.4, 0.15 M sodium chloride and 15 mM sodium azide.

Application

Anti-3,3′,5-Triiodo-L-Thyronine (T3) antibody produced in rabbit was used in flow cytometry analysis of hormone content of rat white blood cells and mast cells.

Biochem/physiol Actions

3,3′,5-Triiodo-L-Thyronine (T3) is the most biologically active form of thyroid hormone that is a key regulator of metabolism, heart, muscle and digestive functions, brain development and maintenance of bone density. The effects of T3 are mediated by its binding to thyroid hormone receptors expressed on human hepatocytes.

Price and Availability


Antibody Explorer
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?