• USA Home
  • T2816 - Clear-view Snap-Cap microtubes
EMAIL THIS PAGE TO A FRIEND
T2816 Sigma
Clear-view Snap-Cap microtubes
size 0.6 mL, green
Purchase
Properties
Related Categories Clear-view snap-cap PCR tubes, Excel Scientific, Excel Scientific Clear-View Snap-Cap Microtubes, Genomics, PCR and Arrays, Labware,
material   cap (Angled cap lid)
  polypropylene
feature   RCF 20,000 × g
  RNase and DNase free
packaging   case of 10,000
mfr. no.   Excel Scientific, MCT-0.6G
size   0.6 mL
color   green
Description
General description
• Superior clarity for sample viewing
• A consistent, ring-lock seal prevents inadvertent opening
• Angled cap lid makes opening easy without thumb fatigue
• Large flat tops for ease of identification and a pierceable cap
• Large, crisp reference lines and graduation numbers
• Certified RNase/DNase free
Legal Information
Clear-view is a trademark of Excel Scientific, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?