• USA Home
  • T2821 - ProteoGel Tris-Tricine-SDS Electrode Buffer

EMAIL THIS PAGE TO A FRIEND
T2821 Sigma

ProteoGel Tris-Tricine-SDS Electrode Buffer

Purchase

Description

Application

SDS-PAGE electrode buffer for Tris-tricine, Tris-glycine, Tris-acetate, or Tris-HCl gels.

Quantity

One pouch makes a total of 5 liters of 1× electrode buffer containing 0.1% SDS, 0.05 M Tris, and 0.05 M tricine, pH approximately 8.2; sold in sets of 10 pouches.

Legal Information

ProteoGel is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Detect Your Unseen Protein

Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?