• USA Home
  • T2906 - Microcentrifuge tubes, extended capacity
EMAIL THIS PAGE TO A FRIEND
T2906 Sigma
Microcentrifuge tubes, extended capacity
volume 1.7 mL , graduated, non-siliconized polypropylene
Purchase
Properties
Related Categories 1.0 to 2.0 mL microcentrifuge tubes, Centrifugation Supplies, Labware, Microcentrifuge Tubes
material   cap
  non-siliconized polypropylene
packaging   case of 500
volume   1.7 mL , graduated
Description
General description
The tubes listed below are specifically designed to hold more volume than other comparable tubes while still fitting all of the most commonly used microcentrifuges. Writing surface on flat lids and side wall for quick sample ID.
The lids provide an easily punctured syringe port for easy access to the contents without opening the tube. Siliconized tubes simplify research requiring low adhesion surface.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Protocols
Enzymatic Assay of Lipase Type XIII from Pseudomonas Species and Pseudomonas Cepacia using a coupled enzyme system of Glycerol Kinase and Glycerophosphate Oxidase (EC 3.1.1.3)
To standardize a procedure for the enzymatic assay of Lipase Type XIII from Pseudomonas Species, Sigma Product Number L9518 , and Pseudomonas Cepacia, Sigma Product Number L9156, using a coupled enzy...
Keywords: Centrifugation, Extinction coefficient, High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?