• USA Home
  • T2920 - Eppendorf® Safe-Lock microcentrifuge tubes
EMAIL THIS PAGE TO A FRIEND
T2920 Sigma
Eppendorf® Safe-Lock microcentrifuge tubes
volume 2.0 mL, amber
Purchase
Properties
Related Categories 1.0 to 2.0 mL microcentrifuge tubes, Centrifugation Supplies, Eppendorf, Eppendorf Safe-Lock Microcentrifuge Tubes, Eppendorf Tubes,
material   cap (Safe-Lock lid)
  polypropylene
feature   RCF 30,000 × g
packaging   case of 1,000
  pack of 500
mfr. no.   Eppendorf®, 0030 120.248 (European)
  Eppendorf®, 022363379 (USA)
volume   2.0 mL
color   amber
Description
Features and Benefits
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?