• USA Home
  • T3034 - Trehalose 6,6′-dimycolate from Mycobacterium bovis

EMAIL THIS PAGE TO A FRIEND
T3034 Sigma

Trehalose 6,6′-dimycolate from Mycobacterium bovis

≥97% 6,6′-mycolate esters basis

Synonym: 6,6′-Di-O-mycoloyl-α,α-trehalose from Mycobacterium bovis, Cord factor from Mycobacterium bovis

Purchase

Properties

Related Categories Bacteria-derived adjuvants, Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z,
assay   ≥97% 6,6′-mycolate esters basis
storage temp.   −20°C

Description

Biochem/physiol Actions

Cord factor is a glycolipid released from Mycobacterium that becomes toxic and antigenic when it interacts with cell surface lipids. It activates macrophages to release inflammatory cytokines and induces thymic atrophy and granuloma formation in lung and other organs. It is responsible for the necrosis and cavitation observed in secondary tuberculosis.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?