• USA Home
  • T3039 - Silicone Rubber Tubing
EMAIL THIS PAGE TO A FRIEND
T3039 Sigma
Silicone Rubber Tubing
Purchase
Description
General description
Natural translucent tubing; exceptionally well suited to use in peristaltic pumps; inert; resilient, elastic, and resistant to compression set; resists UV, moisture; very wide temperature range in use: −75 to 260°C
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?