EMAIL THIS PAGE TO A FRIEND
T3040 Sigma
gel staining Tray
Purchase
Description
Application
For use with aqueous staining and destaining solutions only
Features and Benefits
Constructed of polymethacrylate
• 7.5 in (19 cm) square to fit most gel sizes
• Optically clear down to 300 nm; photograph gels with UV or visible light right in the tray
• Small feet at corners protect bottom from scratching
• Stackable: rock several gels at once; stacking seals tray below to prevent evaporation and spills
• Frosted labeling surface
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
T0567 Gel staining tray, L × W 7 1/2 in. (19 cm) × 7 1/2 in. (19 cm)
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?