• USA Home
  • T3071 - Octakis (2,3,6-tri-O-methyl)-γ-cyclodextrin

EMAIL THIS PAGE TO A FRIEND
T3071 Sigma

Octakis (2,3,6-tri-O-methyl)-γ-cyclodextrin

>97%

  • Empirical Formula (Hill Notation) C72H128O40

  • Molecular Weight 1633.76

  •  MDL number MFCD00210128

  •  PubChem Substance ID 329826735

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Chelation/Complexation Compounds,
InChI Key   JOHZOQCMGQMKML-UCQHMBSKSA-N
assay   >97%
storage temp.   2-8°C

Description

Application

For capillary electrophoresis

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?