• USA Home
  • T3197 - Tissue protein, cell membrane

EMAIL THIS PAGE TO A FRIEND
T3197 Sigma

Tissue protein, cell membrane

from mouse skeletal muscle

Purchase

Properties

Related Categories Antibodies, Cell Biology, Cell Signaling and Neuroscience, Immunohistology and Blotting Controls, Signal Transduction Research Tools,
biological source   from mouse skeletal muscle
form   liquid
shipped in   dry ice
storage temp.   −70°C

Description

Application

Highly purified cell membrane protein from whole tissue homogenates. Useful in electrophoresis, Western blotting, immunoassay, and other applications requiring source-specific, non-denatured protein.

Physical form

0.5 mg protein (approx. 3 mg protein/mL)

Supplied in a solution containing 20 mM HEPES, 1.5 mM MgCl2, 10 mM KCl, 40 mM sucrose, 1 mM EDTA, 2% glycerol, 0.5% sodium deoxycholate, 1% NP-40, pH 7.9, and a cocktail of protease inhibitors.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?