• USA Home
  • T3281 - Microcentrifuge tubes, extended capacity
EMAIL THIS PAGE TO A FRIEND
T3281 Sigma
Microcentrifuge tubes, extended capacity
volume 0.65 mL , graduated, siliconized polypropylene
  •  eCl@ss 32119210
Purchase
Properties
Related Categories 0.4 to 0.7 mL microcentrifuge tubes, Centrifugation Supplies, Labware, Microcentrifuge Tubes
material   cap
  siliconized polypropylene
packaging   pack of 500 ea
volume   0.65 mL , graduated
Description
General description
The tubes listed below are specifically designed to hold more volume than other comparable tubes while still fitting all of the most commonly used microcentrifuges. Writing surface on flat lids and side wall for quick sample ID.
The lids provide an easily punctured syringe port for easy access to the contents without opening the tube. Siliconized tubes simplify research requiring low adhesion surface.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?