• USA Home
  • T3290 - 4-Trifluoromethylumbelliferone 7-acetate

EMAIL THIS PAGE TO A FRIEND
T3290 Sigma

4-Trifluoromethylumbelliferone 7-acetate

~99% (TLC)

Purchase

Properties

assay   ~99% (TLC)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?