• USA Home
  • T3640 - Tropomyosin from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
T3640 Sigma

Tropomyosin from rabbit muscle

lyophilized powder

Purchase

Properties

form   lyophilized powder
storage temp.   −20°C

Description

Analysis Note

SDS electrophoresis shows two major bands.

Packaging

Package size based on protein content

Physical form

Lyophilized powder; may contain Tris buffer salts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?