EMAIL THIS PAGE TO A FRIEND
T3695 Sigma

TFLZn-AM

≥90% (TLC)

Synonym: 4-[([6-Methoxy-2-methyl-8-quinolinyl]amino)sulfonyl]benzoic acid acetoxymethyl ester

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes, T-U-V More...
InChI Key   ZCVIWNFCXWULOJ-UHFFFAOYSA-N
assay   ≥90% (TLC)
color   pale yellow
storage temp.   −20°C

Description

Analysis Note

The TFLZn-Zn complex is fluorescent, with an excitation at 380 nm and emission at 510 nm.

Biochem/physiol Actions

Cell-permeant derivative of the intracellular Zn++ indicator

Caution

Protect from light

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?