• USA Home
  • T3696 - Hexakis (2,3,6-tri-O-acetyl)-α-cyclodextrin

EMAIL THIS PAGE TO A FRIEND
T3696 Sigma

Hexakis (2,3,6-tri-O-acetyl)-α-cyclodextrin

>98%

  • Empirical Formula (Hill Notation) C72H96O48

  • Molecular Weight 1729.50

  •  MDL number MFCD03453025

  •  PubChem Substance ID 329826774

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Chelation/Complexation Compounds,
InChI Key   WPCDNMKUJVKODC-WLPBKHADSA-N
assay   >98%
storage temp.   2-8°C

Description

Application

For capillary electrophoresis, chiral selector

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?