• USA Home
  • T3797 - Trichosanthin from Trichosanthes kirilowii

EMAIL THIS PAGE TO A FRIEND
T3797 Sigma

Trichosanthin from Trichosanthes kirilowii

~95% (SDS-PAGE), lyophilized powder

Purchase

Properties

assay   ~95% (SDS-PAGE)
form   lyophilized powder
mol wt   mol wt 25 kDa
composition   Protein, ~15% Lowry
storage temp.   −20°C

Description

Biochem/physiol Actions

Active principle of the traditional Chinese medicinal plant with abortifacient activity.1 It is a potent protein synthesis inhibitor in vitro1,2 and an inhibitor of HIV replication.3

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sodium phosphate and preservatives

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
YD0806000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?