• USA Home
  • T3818 - Troubleshooting HPLC Systems: A Bench Manual
EMAIL THIS PAGE TO A FRIEND
T3818 Sigma
Troubleshooting HPLC Systems: A Bench Manual
  •  ISBN-10 0-471-17834-9
  •  ISBN-13 978-0-471-17834-7
Purchase
Properties
Related Categories Books and Software, Chromatography Books, Labware More...
publication info   P.C. Sadek, John Wiley & Sons, 1999, 320 pp., soft cover
mfr. no.   Wiley
Description
General description
This book provides a useful guide to the new generation of HPLC equipment and techniques. With emphasis on effective troubleshooting of HPLC systems, it covers system configuration and functions, problem-solving procedures, maintenance, and HPLC basics. It then guides users investigating the source of a malfunction through each system component-from solvents and reservoirs to sample preparation to columns and detectors. Special features include a detailed review of HPLC instrumentation and accessories, role of operating parameters as indicators of system performance, and tables and charts detailing solvents′ properties.
Table of Contents
HPLC System Components.
Fundamental Terminology, Parameters, Variables, and Theory.
Troubleshooting by HPLC Component.
Glossary.
Appendix.
Quick Troubleshooting Guide.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?