• USA Home
  • T3942 - Eppendorf® exchangeable thermoblocks, with lid
EMAIL THIS PAGE TO A FRIEND
T3942 Sigma
Eppendorf® exchangeable thermoblocks, with lid
thermoblock for microtiter plates (MTPs) and deepwell plates, accepts 1 x 96 well microplate, w/ lid, 1/cs
Synonym: PCR cooling block, PCR tube rack, cool block, cooling block, heat block, heating block
Purchase
Properties
Related Categories Auxiliary Equipment, Block Heaters, Eppendorf, Heating, HybriScan Rapid Test Systems for Food & Beverages,
feature   lid
packaging   case of 1
  pack of 1
mfr. no.   Eppendorf®, 022670565 (USA)
  Eppendorf®, 5363 000.012 (European)
W × D × H   135 mm × 90 mm × 60 mm
Description
General description
Eppendorf® Exchangeable Thermoblocks
The exchangeable thermoblock system for the Thermomixer Comfort (T3317) and ThermoStat Plus ensure a precise "fit" for all common lab tubes: From simpls temperature control of a chemical to complex enzymatic reactions and hybridization experiments, and from microtiter plates (MTPs) to 50 mL conical tubes- the application possibilities are practically endless.
Suitable for Eppendorf PCR plate adapters (A7707)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
HybriScan™ Rapid Test Systems: Rapid detection, identification and quantification of microorganisms in beverages, food and water
The new HybriScan Test System, which uses sandwich hybridization, provides fast, sensitive and reliable detection, identification and quantification of spoilage and pathogenic microorganisms in bever...
Jvo Siegrist
AnalytiX Volume 8 Article 3
Keywords: Enzyme-linked immunosorbent assay, Microbiology, Nucleic acid hybridization, Polymerase chain reaction, Purification, Transcription
Microbiology Introduction
An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins
Testing EZ-Fit™ Products Family for Bioburden Testing
The EZ family of products has been designed to optimize and streamline the bioburden workflow. The family includes:
Keywords: Filtration, Separation
Related Content
HybriScan Rapid Test Systems for Microorganisms in Food & Beverages
- Introduction - Benefits over conventional detection methods and PCR - Table with advantages over other detection techniques - Principles of the HybriScan method - Sensitivity, specificity, flexibil...
Keywords: Cell disruption, Clinical, Detection methods, Enzyme-linked immunosorbent assay, Food Safety, Nucleic acid hybridization, Polymerase chain reaction, Purification, Transcription
HybriScan Rapid Test Systems
Das neue HybriScan Testsystem, welches auf der Sandwich-Hybridisierung basiert, bietet einen schnellen, empfindlichen und zuverlässigen Weg Verderbsmikroorganismen und pathogenen Keime in Getränke, N...
Keywords: Cell disruption, Enzyme-linked immunosorbent assay, Nucleic acid hybridization, Polymerase chain reaction, Transcription
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?