• USA Home
  • T4327 - Monoclonal Anti-Tom22−Atto 488 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
T4327 Sigma

Monoclonal Anti-Tom22−Atto 488 antibody produced in mouse

1.5-3.0 mg/mL, clone IC9-2, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-Mitochondrial import receptor Tom22, Anti-TOMM22, Anti-Translocase of outer mitochondrial membrane 22 homolog (yeast), Monoclonal Anti-TOM22

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Mitochondrial Proteins,
species reactivity   monkey, human
does not react with   rodent
application(s)   direct immunofluorescence: 1-2 μg/mL using human HeLa cells
clone   IC9-2, monoclonal
concentration   1.5-3.0 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 22 kDa
shipped in   dry ice
storage condition   protect from light
storage temp.   −20°C
Gene Information   human ... TOMM22(56993)
biological source   mouse
conjugate   Atto 488 conjugate

Description

Immunogen

membrane fraction from Vero (monkey kidney-derived) cells.

General description

find more information here

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence

Durapore Membrane Filters
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

T6319 Monoclonal Anti-TOM22 antibody produced in mouse, ~1 mg/mL, clone 1C9-2, purified immunoglobulin, buffered aqueous solution
HPA003037 Anti-TOMM22 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?