• USA Home
  • T4398 - Thyroglobulin−Agarose

EMAIL THIS PAGE TO A FRIEND
T4398 Sigma

Thyroglobulin−Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Specialty Resins More...
biological source   Thyroglobulin Porcine
form   saline suspension
matrix   Cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix spacer   1 atom
capacity   0.5-2 mg/mL binding capacity (Phaseolus vulgaris agglutinin)
storage temp.   2-8°C

Description

Physical form

Suspension in 1.0 M NaCl containing 0.02% thimerosal

Application

Thyroglobulin-agarose is use in protein chromatography, affinity chromatography and specialty resins. Thyroglobulin-agarose has been used to study apoptosis of concanavalin A-stimulated mouse lymphocytes. Thyroglobulin-agarose has also been used in a study to determine that xenotransplantation of porcine organs into HLA-sensitized patients awaiting kidney transplantation has the potential to be rejected by humoral mechanisms, thereby recommending testing to avoid such cross-reactive antibodies.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?