• USA Home
  • T4434 - Trypsin TPCK treated from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
T4434 Sigma

Trypsin TPCK treated from bovine pancreas

Purchase

Properties

storage temp.   −20°C

Description

Analysis Note

Protein determined by E1%/280 unless otherwise indicated.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH_7.6 at 25° C using BAEE as substrate. Reaction volume = 3.2_ml (1_cm light path).

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
YN5075000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?