EMAIL THIS PAGE TO A FRIEND
T4550 Sigma

Tata binding protein

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ~50 fpu/μg protein (Bradford)

Purchase

Properties

recombinant   expressed in E. coli
assay   ≥95% (SDS-PAGE)
shipped in   dry ice
storage temp.   −70°C

Description

Packaging

Package size based on protein content

Unit Definition

One fpu (footprint unit) fully protects the TATA box of the adenovirus mlp in a DNase I footprint assay.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?