EMAIL THIS PAGE TO A FRIEND
T4645 Sigma

Trityl−Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Hydrophobic Interaction, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   50-100 μmol per mL
matrix   Cross-linked 4% beaded agarose
matrix attachment   direct
storage temp.   2-8°C

Description

Physical form

Suspension in 1.0 M NaCl

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?