• USA Home
  • T4770 - Tropomyosin from bovine muscle

EMAIL THIS PAGE TO A FRIEND
T4770 Sigma

Tropomyosin from bovine muscle

lyophilized powder (contains Tris buffer salts)

Purchase

Properties

form   lyophilized powder (contains Tris buffer salts)
storage temp.   −20°C
Gene Information   cow ... TPM1(281544), TPM2(497015), TPM3(497019)

Description

Analysis Note

SDS electrophoresis shows two major bands.

Biochem/physiol Actions

In the absence of Ca++, inhibits muscle contractility by blocking the myosin binding sites on actin.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?