• USA Home
  • T4817 - Anti-Transforming Growth Factor-β5 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
T4817 Sigma

Anti-Transforming Growth Factor-β5 antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym: Anti-TGF-β5

Purchase

Description

Immunogen

recombinant amphibian transforming growth factor-β5 expressed in Sf 21 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Application

Anti-TGF-β5 antibody is suitable for neutralization reactions (ND50 is 0.03-0.08 μg/ml). The antibody may be used for immunoblotting at a working concentration of 1 μg/ml. For ELISA a working concentration of 1 μg/ml is recommended.

General description

Transforming growth factor-β5 (TGF-β) belongs to the TGF-β superfamily of secreted proteins. The TGF-β family includes five isoforms, TGF-β1 toTGF-β5. The isoforms are involved in a variety of signaling pathways that involve serine/threonine protein kinases. TGF-β5 isoform is expressed in the embryonic as well as adult stages of Xenopus. TGF-β5 is involved in cell and tissue differentiation and proliferation. TGF-β5 expression is not observed in mammals
Anti-TGF-β5 recognizes the amphibian TGF-β5. It does not show cross reactivity with TGF-β1, porcine TGF-β2, TGF-β2, or TGF-β3.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?