• USA Home
  • T4819 - Tempoxime hydrochloride

EMAIL THIS PAGE TO A FRIEND
T4819 Sigma

Tempoxime hydrochloride

≥98% (TLC)

Synonym: 2,2,6,6-tetramethypiperidone-4 oxime hydrochloride, TMP=NOH

  • Empirical Formula (Hill Notation) C9H18N2O · HCl

  • Molecular Weight 206.71

  •  MDL number MFCD00160775

  •  PubChem Substance ID 24900229

Purchase

Properties

Related Categories Antagonists, Cell Biology, Cell Signaling and Neuroscience, Cholinergics, Ion Channels,
InChI Key   PMDFGKBTEGXYED-UHFFFAOYSA-N
assay   ≥98% (TLC)
solubility   H2O: soluble

Description

Biochem/physiol Actions

Neuronal nicotinic AChR antagonist and potent ganglioblocker.

Preparation Note

Tempoxime hydrochloride is soluble in water.

Physical form

Colorless crystal powder

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?