EMAIL THIS PAGE TO A FRIEND
T4888 Sigma

Trimethyl phosphite

~97%

Purchase

Properties

assay   ~97%
density   1.05 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2329 3 / PGIII
WGK Germany 
1
RTECS 
TH1400000
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T79707 Trimethyl phosphite, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?