EMAIL THIS PAGE TO A FRIEND
T4905 Sigma

Tri-O-thymotide

Synonym: Cyclic trilactone of o-thymotic acid

Purchase

Properties

InChI Key   ZIKHLMVJGCYVAC-UHFFFAOYSA-N
mp   219-221 °C(lit.)

Description

Biochem/physiol Actions

Host substance for formation of clathrates (inclusion complexes).

Linkage

Cyclic trilactone of thymotic acid (3-isopropylsalicylic acid).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?