EMAIL THIS PAGE TO A FRIEND
T5068 Sigma

Tyrphostin AG 658

≥98%, solid

Synonym: 4-Hydroxy-3-methoxy-5-(phenylthiomethyl)benzylidinecyanoacetamide, Tyrphostin C6

  • Empirical Formula (Hill Notation) C18H16N2O3S

  • Molecular Weight 340.40

  •  MDL number MFCD00673898

  •  PubChem Substance ID 329826806

Purchase

Description

Biochem/physiol Actions

EGFR protein tyrosine kinase inhibitor.

Preparation Note

Tyrphostin AG 658 is soluble in DMSO and methanol.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?