EMAIL THIS PAGE TO A FRIEND
T5318 Sigma

Tyrphostin AG 808

≥98%

Synonym: 2-Cyano-3-(3′,4′-dihydroxyphenyl)-1-(3″-indolyl)-3-oxo-1-propene

  • Empirical Formula (Hill Notation) C18H12N2O3

  • Molecular Weight 304.30

  •  MDL number MFCD00906779

  •  PubChem Substance ID 24278744

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Kinase/Phosphatase Biology, Protein Kinase Inhibitors
InChI Key   TTZDOBVWBQXGDC-KPKJPENVSA-N
assay   ≥98%
solubility   DMSO: soluble
storage temp.   −20°C
Gene Information   human ... MST1R(4486), PTK2(5747), PTK6(5753), PTK7(5754), TWF1(5756), TXK(7294)
mouse ... Egfr(13649), Pdgfrb(18596)

Description

Biochem/physiol Actions

Protein tyrosine kinase inhibitor

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?