EMAIL THIS PAGE TO A FRIEND
T5379 Sigma

Tyr-Phe

  • CAS Number 17355-11-2

  • Empirical Formula (Hill Notation) C18H20N2O4

  • Molecular Weight 328.36

  •  MDL number MFCD00037255

  •  PubChem Substance ID 24900270

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   CGWAPUBOXJWXMS-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?