• USA Home
  • T5517 - Tyr-D-Trp-Ala-Trp-D-Phe amide

EMAIL THIS PAGE TO A FRIEND
T5517 Sigma

Tyr-D-Trp-Ala-Trp-D-Phe amide

≥95% (HPLC)

Synonym: Momany peptide

  • CAS Number 76338-79-9

  • Empirical Formula (Hill Notation) C43H46N8O6

  • Molecular Weight 770.88

  •  MDL number MFCD00133893

  •  PubChem Substance ID 24900291

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   JFMKMZZAVDKJAH-UHFFFAOYSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Releases growth hormone from rat pituitary in vitro.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?