• USA Home
  • T5538 - C-Flex® laboratory tubing
EMAIL THIS PAGE TO A FRIEND
T5538 Sigma
C-Flex® laboratory tubing
I.D. × O.D. 5/16 in. × 9/16 in.
Purchase
Properties
I.D. × O.D.   5/16 in. × 9/16 in.
valve size   1/8 in.
Description
General description
Tubing is opaque, white thermoplastic elastomer formulated from styrene-ethylene-butadiene-styrene (SEBS) block polymer. Cost-effective alternative to silicone in many applications including peristaltic pumps. Temp. range: -100 to 100°C.
Legal Information
C-Flex is a registered trademark of Concept Polymer Technologies Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?