EMAIL THIS PAGE TO A FRIEND
T5549 Sigma

Thiourea-14C

solid

Purchase

Properties

InChI Key   UMGDCJDMYOKAJW-NJFSPNSNSA-N
form   solid
extent of labeling   1-30 mCi per mmol
mass shift   M+2
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?