• USA Home
  • T5791 - Glass tubes for tube gels
EMAIL THIS PAGE TO A FRIEND
T5791 Sigma
Glass tubes for tube gels
I.D. × L 1.0 mm × 13.5 cm
Purchase
Description
Compatibility
The 7.5 cm tubes fit the mini electrophoresis unit, and the 13.5 cm tubes fit the standard-size unit.
Features and Benefits
Although the tubes listed below have different inside diameters, they all have the same 3.0 mm outside diameter. This makes the tubes less fragile and easier to handle.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?