• USA Home
  • T5901 - Thymus acetone powder from calf

EMAIL THIS PAGE TO A FRIEND
T5901 Sigma

Thymus acetone powder from calf

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

T6138 Thymus acetone powder from rabbit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?