• USA Home
  • T5909 - Glyceryl tripalmitelaidate

EMAIL THIS PAGE TO A FRIEND
T5909 Sigma

Glyceryl tripalmitelaidate

≥99%

Synonym: 1,2,3-tri-(9Z-hexadecenoyl)-glycerol, TG(16:1(9Z)/16:1(9Z)/16:1(9Z)), Tripalmitelaidin

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerolipid Metabolism, Glycerolipids, Lipids,
InChI Key   SKGWNZXOCSYJQL-IKVQWSBMSA-N
assay   ≥99%
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?