• USA Home
  • T5914 - Silicone Rubber Tubing
EMAIL THIS PAGE TO A FRIEND
T5914 Sigma
Silicone Rubber Tubing
Purchase
Properties
Related Categories Cell Biology, Dissociation, General Stem Cell Biology, Stem Cell Biology, Stem Cell Isolation More...
I.D. × wall × O.D.   3/4 in. (19.0 mm) × 3/16 in. (4.8 mm) × 1 1/8 in. (28.6 mm)
Description
General description
Natural translucent tubing; exceptionally well suited to use in peristaltic pumps; inert; resilient, elastic, and resistant to compression set; resists UV, moisture; very wide temperature range in use: −75 to 260°C
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?