• USA Home
  • T6004 - Tautomerase from bovine kidney

EMAIL THIS PAGE TO A FRIEND
T6004 Sigma

Tautomerase from bovine kidney

Grade I, aqueous solution, 1-4 units/mg protein (Lowry), ~10 units/mL

Synonym: Phenylpyruvate keto-enol isomerase

Purchase

Properties

Related Categories 5.3.x.x Intramolecular oxidoreductases, 5.x.x.x Isomerases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
type   Grade I
form   aqueous solution
concentration   ~10 units/mL
shipped in   wet ice
storage temp.   −20°C

Description

Unit Definition

One unit will produce a first-order rate constant (k) of 1.0 at pH 6.2 at 25 °C, using p-hydroxyphenylpyruvate (keto). Reaction vol = 3.3 ml.

Application

Tautomerase from bovine kidney has been used in a study to assess tritium isotope effects in the reaction catalyzed by 4-hydroxyphenylpyruvate dioxygenase. Tautomerase from bovine kidney has also been used in a study to investigate human macrophage migration inhibitory factor.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?