• USA Home
  • T6013 - apo-Transferrin from rat

EMAIL THIS PAGE TO A FRIEND
T6013 Sigma

apo-Transferrin from rat

~98%

Purchase

Properties

assay   ~98%
solubility   H2O: 20 mg/mL
storage temp.   2-8°C
Gene Information   rat ... Tf(24825)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

T0523 apo-Transferrin from mouse, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?