• USA Home
  • T6037 - Tetrahydrocortisone 3-(β-D-glucuronide)

EMAIL THIS PAGE TO A FRIEND
T6037 Sigma

Tetrahydrocortisone 3-(β-D-glucuronide)

Synonym: 5β-Pregnane-3α,17α,21-triol-11,20-dione 3-(β-D-glucuronide)

Purchase

Properties

form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?